KEY195583071 Key Organics / BIONET

4-[5-(2-methylpropyl)-1,2,4-oxadiazol-3-yl]-2-(piperazin-1-yl)pyrimidine dihydrochloride

  • MDL number MFCD12025835
  • Cas Number : 1177093-07-0
  • Molecular Weight : 361.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94519

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0122_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart