KEY195583072 Key Organics / BIONET

4-(5-cyclopropyl-1,2,4-oxadiazol-3-yl)-2-(piperazin-1-yl)pyrimidine dihydrochloride

  • MDL number MFCD12025836
  • Cas Number : 1177093-00-3
  • Molecular Weight : 345.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94522

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0179_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart