KEY195583073 Key Organics / BIONET

2-(piperazin-1-yl)-4-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyrimidine dihydrochloride

  • MDL number MFCD12025837
  • Cas Number : 1177093-00-3
  • Molecular Weight : 373.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94524

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0194_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart