KEY195583074 Key Organics / BIONET

2-chloro-5-fluoro-6-methoxypyridine-3-carboxylic acid

  • MDL number MFCD12025843
  • Cas Number : 943025-86-3
  • Molecular Weight : 205.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164 - 166
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94576

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0759_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart