KEY195583075 Key Organics / BIONET

(E)-(2,2-dimethoxyethyl)(phenylmethylidene)amine

  • MDL number MFCD08056680
  • Cas Number : 54879-74-2
  • Molecular Weight : 193.246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94815

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-0002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart