KEY195583095 Key Organics / BIONET

(2,5-difluoro-4-nitrophenyl)methanol

  • MDL number MFCD09909606
  • Cas Number : 1160474-68-9
  • Molecular Weight : 189.118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 81
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96124

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DD-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart