KEY195583100 Key Organics / BIONET

methyl 5-chloro-6-methoxypyridine-3-carboxylate

  • MDL number MFCD12025914
  • Cas Number : 220656-93-9
  • Molecular Weight : 201.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 110
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96133

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DD-0729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart