KEY195583105 Key Organics / BIONET

[4-(1,3-oxazol-5-yl)phenyl]methanol

  • MDL number MFCD02682042
  • Cas Number : 179057-18-2
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 107 - 109
Iupac Name [4-(1,3-oxazol-5-yl)phenyl]methanol
Inchi Code 1S/C10H9NO2/c12-6-8-1-3-9(4-2-8)10-5-11-7-13-10/h1-5,7,12H,6H2
Inchi Key KUGBYBXRZICBQU-UHFFFAOYSA-N

Description

Melting Point 107 - 109
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96142

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DD-0740_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart