KEY195583128 Key Organics / BIONET

3-amino-4-fluorobenzenesulfonic acid

  • MDL number MFCD12025945
  • Cas Number : 349-64-4
  • Molecular Weight : 191.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108220

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3000_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart