KEY195583145 Key Organics / BIONET

6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine

  • MDL number MFCD04619561
  • Cas Number : 189116-20-9
  • Molecular Weight : 220.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 122 - 124
Iupac Name 6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Inchi Code 1S/C8H4ClF3N2/c9-5-1-2-7-13-6(8(10,11)12)4-14(7)3-5/h1-4H
Inchi Key NRZYRLQVLCUDQH-UHFFFAOYSA-N

Description

Melting Point 122 - 124
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96403

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/ED-0229_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart