KEY195583155 Key Organics / BIONET

3-amino-1,2,3,4-tetrahydroquinolin-2-one hydrochloride

  • MDL number MFCD09999258
  • Cas Number : 35849-31-1
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 314 - 317 (dec)
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96119

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DD-0708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart