KEY195583165 Key Organics / BIONET

3-bromo-2-methyl-6-nitro-2H-indazole

  • MDL number MFCD02666041
  • Cas Number : 74209-41-9
  • Molecular Weight : 256.059

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174 - 176
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96434

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ED-0736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart