KEY195583167 Key Organics / BIONET

methyl 2,5,6-trichloropyrimidine-4-carboxylate

  • MDL number MFCD12025876
  • Cas Number : 89284-85-5
  • Molecular Weight : 241.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46 - 49
Purity 97
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95660

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CD-0064_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart