KEY195583182 Key Organics / BIONET

hexahydro-1H-pyrrolo[1,2-c]imidazole-1,3-dione

  • MDL number MFCD01851016
  • Cas Number : 5768-79-6
  • Molecular Weight : 140.142

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 162
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101751

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LC-0098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart