KEY195583186 Key Organics / BIONET

2-[1,1'-biphenyl]-4-yl-1-ethanol

  • MDL number MFCD09926182
  • Cas Number : 37729-18-3
  • Molecular Weight : 198.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108218

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2998_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart