KEY195583188 Key Organics / BIONET

6-bromo-1H,2H,4H-pyrido[2,3-d][1,3]oxazin-2-one

  • MDL number MFCD12025916
  • Cas Number : 335032-38-7
  • Molecular Weight : 229.033

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237 - 239
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96135

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DD-0732_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart