KEY195583192 Key Organics / BIONET

methyl 2-(4-amino-3-iodophenyl)-2-methylpropanoate

  • MDL number MFCD12025947
  • Cas Number : 756813-88-4
  • Molecular Weight : 319.142

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108230

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart