KEY235883501 Key Organics / BIONET

tert-butyl 8-benzyl-2,8-diazaspiro[4.5]decane-2-carboxylate

  • MDL number MFCD12400945
  • Cas Number : 336191-16-3
  • Molecular Weight : 330.472

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76736

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1515_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart