KEY235883516 Key Organics / BIONET

methyl 5-(trifluoromethoxy)-1H-indazole-3-carboxylate

  • MDL number MFCD12498703
  • Cas Number : 932041-12-8
  • Molecular Weight : 260.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181 - 182
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96866

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FD-0068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart