KEY235883522 Key Organics / BIONET

6-chloro-7-iodo-1H-pyrrolo[3,2-c]pyridine

  • MDL number MFCD12498711
  • Cas Number : 1171919-03-1
  • Molecular Weight : 278.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 245
Iupac Name 6-chloro-7-iodo-1H-pyrrolo[3,2-c]pyridine
Inchi Code 1S/C7H4ClIN2/c8-7-5(9)6-4(3-11-7)1-2-10-6/h1-3,10H
Inchi Key KBQWSFZIEJULAZ-UHFFFAOYSA-N

Description

Melting Point 245
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96898

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FD-0711_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart