KEY235883540 Key Organics / BIONET

4-chloropyridine-2,6-dicarboxylic acid

  • MDL number MFCD04117894
  • Cas Number : 4722-94-5
  • Molecular Weight : 201.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp dec 210
Iupac Name 4-chloro-2,6-pyridinedicarboxylic acid
Inchi Code 1S/C7H4ClNO4/c8-3-1-4(6(10)11)9-5(2-3)7(12)13/h1-2H,(H,10,11)(H,12,13)
Inchi Key IYUMNONNHYADBU-UHFFFAOYSA-N

Description

Melting Point dec 210
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96413

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/ED-0255_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart