KEY235883543 Key Organics / BIONET

5-methyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazol-4-ol

  • MDL number MFCD03426594
  • Cas Number : 119868-25-6
  • Molecular Weight : 242.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85148

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7G-317S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart