KEY235883544 Key Organics / BIONET

[2-(piperidin-1-yl)phenyl]methanol

  • MDL number MFCD05864821
  • Cas Number : 87066-94-2
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(1-piperidinyl)phenyl]methanol
Inchi Code 1S/C12H17NO/c14-10-11-6-2-3-7-12(11)13-8-4-1-5-9-13/h2-3,6-7,14H,1,4-5,8-10H2
Inchi Key XWFXVEQCVPXALT-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96444

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/ED-0758_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart