KEY235883550 Key Organics / BIONET

6-amino-2-chloropyrimidine-4-carboxamide

  • MDL number MFCD12498714
  • Cas Number : 500352-09-0
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96909

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FD-0729_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart