KEY235883558 Key Organics / BIONET

2-(hydroxymethyl)-5-methoxy-4H-pyran-4-one

  • MDL number MFCD00224874
  • Cas Number : 6269-25-6
  • Molecular Weight : 156.137

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 167
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96864

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FD-0057_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart