KEY235883565 Key Organics / BIONET

2,4,6-trichlorophenyl 2-cyanobenzene-1-sulfonate

  • MDL number MFCD12498725
  • Cas Number : 1171919-29-1
  • Molecular Weight : 362.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 181
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106623

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/NA-0219_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart