KEY235883566 Key Organics / BIONET

2,4,6-trichlorophenyl 5-bromothiophene-2-sulfonate

  • MDL number MFCD12498726
  • Cas Number : 1171919-31-5
  • Molecular Weight : 422.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54 - 56
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106624

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NA-0226_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart