KEY235883569 Key Organics / BIONET

2,4,6-trichlorophenyl 2-fluorobenzene-1-sulfonate

  • MDL number MFCD12498728
  • Cas Number : 1171919-36-0
  • Molecular Weight : 355.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91 - 93
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106627

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NA-0233_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart