KEY235883575 Key Organics / BIONET

2-(hydroxymethyl)-5-methoxy-1,4-dihydropyridin-4-one

  • MDL number MFCD04122652
  • Cas Number : 6323-21-3
  • Molecular Weight : 155.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(hydroxymethyl)-5-methoxy-4-pyridinol
Inchi Code 1S/C7H9NO3/c1-11-7-3-8-5(4-9)2-6(7)10/h2-3,9H,4H2,1H3,(H,8,10)
Inchi Key LEQMEXDHGCSIGW-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96869

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FD-0078_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart