KEY238899214 Key Organics / BIONET

2-amino-5-iodo-3-methylbenzoic acid

  • MDL number MFCD09029816
  • Cas Number : 108857-24-5
  • Molecular Weight : 277.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 235 - 240
Iupac Name 2-amino-5-iodo-3-methylbenzoic acid
Inchi Code 1S/C8H8INO2/c1-4-2-5(9)3-6(7(4)10)8(11)12/h2-3H,10H2,1H3,(H,11,12)
Inchi Key WFKOJKHXWWNXPK-UHFFFAOYSA-N

Description

Melting Point 235 - 240
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97792

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HD-0722_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart