KEY238899216 Key Organics / BIONET

methyl 4-(methylamino)-3-nitrobenzoate

  • MDL number MFCD09862450
  • Cas Number : 36242-50-9
  • Molecular Weight : 210.189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97800

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HD-0736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart