KEY238899301 Key Organics / BIONET

5-acetyl-3-bromo-6-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD12827806
  • Cas Number : 136117-46-9
  • Molecular Weight : 230.061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97408

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GD-0241_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart