KEY238899303 Key Organics / BIONET

3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD12827808
  • Cas Number : 1186405-07-1
  • Molecular Weight : 275.143

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 dec
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97417

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GD-0717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart