KEY238899306 Key Organics / BIONET

methyl 2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetate

  • MDL number MFCD12827811
  • Cas Number : 91640-73-2
  • Molecular Weight : 7/23/1900 6:10:05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97424

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GD-0737_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart