KEY238899312 Key Organics / BIONET

3-oxo-3-{thieno[2,3-b]pyridin-2-yl}propanenitrile

  • MDL number MFCD12827817
  • Cas Number : 1186405-15-1
  • Molecular Weight : 202.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204 - 206
Purity 95
Storage temp. RT
Physical Form Brown to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97434

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GD-0761_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart