KEY238899325 Key Organics / BIONET

1-phenyl-3-(trifluoromethyl)-1H-pyrazol-4-ol

  • MDL number MFCD02327419
  • Cas Number : 119868-24-5
  • Molecular Weight : 228.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116 - 118
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97410

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GD-0702_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart