KEY238899333 Key Organics / BIONET

ethyl thieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD07772793
  • Cas Number : 59944-78-4
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61 - 63
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97421

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GD-0724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart