KEY238899351 Key Organics / BIONET

4-nitro-2-(trifluoromethyl)phenol

  • MDL number MFCD11100972
  • Cas Number : 1548-61-4
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 105 - 107
Iupac Name 4-nitro-2-(trifluoromethyl)phenol
Inchi Code 1S/C7H4F3NO3/c8-7(9,10)5-3-4(11(13)14)1-2-6(5)12/h1-3,12H
Inchi Key GWGZFNRFNIXCGH-UHFFFAOYSA-N

Description

Melting Point 105 - 107
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96873

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/FD-0093_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart