KEY238899352 Key Organics / BIONET

2,5-dichloro-3-(trifluoromethyl)pyridine

  • MDL number MFCD00042244
  • Cas Number : 70158-59-7
  • Molecular Weight : 215.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 82 / 26 mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97774

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HD-0213_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart