KEY238899353 Key Organics / BIONET

ethyl 3-(pyridin-2-yl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD12827769
  • Cas Number : 174139-65-2
  • Molecular Weight : 217.228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 98
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81132

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Z-0611_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart