KEY238899355 Key Organics / BIONET

2-bromo-5,5-dimethylcyclohexane-1,3-dione

  • MDL number MFCD00092415
  • Cas Number : 1195-91-1
  • Molecular Weight : 219.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 158 - 160
Iupac Name 2-bromo-3-hydroxy-5,5-dimethyl-2-cyclohexen-1-one
Inchi Code 1S/C8H11BrO2/c1-8(2)3-5(10)7(9)6(11)4-8/h7H,3-4H2,1-2H3
Inchi Key OCXANUSFMRALNG-UHFFFAOYSA-N

Description

Melting Point 158 - 160
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97780

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/HD-0700_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart