KEY238899356 Key Organics / BIONET

2-bromocyclohexane-1,3-dione

  • MDL number MFCD00971951
  • Cas Number : 60060-44-8
  • Molecular Weight : 191.024

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159 - 161
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97781

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HD-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart