KEY238899364 Key Organics / BIONET

ethyl 4,6-dihydroxy-5-nitropyridine-3-carboxylate

  • MDL number MFCD01765414
  • Cas Number : 6317-97-1
  • Molecular Weight : 228.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 243 - 246
Iupac Name ethyl 6-hydroxy-5-nitro-4-oxo-1,4-dihydro-3-pyridinecarboxylate
Inchi Code 1S/C8H8N2O6/c1-2-16-8(13)4-3-9-7(12)5(6(4)11)10(14)15/h3H,2H2,1H3,(H2,9,11,12)
Inchi Key JBTWWCUXSHUDEJ-UHFFFAOYSA-N

Description

Melting Point 243 - 246
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97790

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HD-0719_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart