KEY240035464 Key Organics / BIONET

methyl 6-bromo-4-oxo-1,4-dihydroquinoline-8-carboxylate

  • MDL number MFCD12922675
  • Cas Number : 1190198-14-1
  • Molecular Weight : 282.093

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220 - 222
Purity 95
Storage temp. RT
Physical Form Off-white to Grey Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96852

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FD-0006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart