KEY240035504 Key Organics / BIONET

6-chloro-4-methoxypyridine-3-carboxylic acid

  • MDL number MFCD10000638
  • Cas Number : 716362-10-6
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101002

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KD-0738_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart