KEY240035508 Key Organics / BIONET

2-(benzyloxy)-4-methoxybenzaldehyde

  • MDL number MFCD02113682
  • Cas Number : 32884-23-4
  • Molecular Weight : 242.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68 - 70
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97772

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HD-0097_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart