KEY240035514 Key Organics / BIONET

5-(benzyloxy)-2-(hydroxymethyl)-4H-pyran-4-one

  • MDL number MFCD01927596
  • Cas Number : 15771-06-9
  • Molecular Weight : 232.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96389

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ED-0040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart