KEY240035518 Key Organics / BIONET

5-(benzyloxy)-2-(hydroxymethyl)pyridin-4-ol

  • MDL number MFCD12922670
  • Cas Number : 165948-37-8
  • Molecular Weight : 231.251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228 - 230
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96391

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ED-0044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart