KEY240035525 Key Organics / BIONET

5-amino-3-methyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD02020550
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 161 - 163
Iupac Name 5-amino-3-methyl-1H-pyrazole-4-carbonitrile
Inchi Code 1S/C5H6N4/c1-3-4(2-6)5(7)9-8-3/h1H3,(H3,7,8,9)
Inchi Key CRRHJAMDQMQGKG-UHFFFAOYSA-N

Description

Melting Point 161 - 163
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100995

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KD-0730_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart