KEY240035529 Key Organics / BIONET

5-chloro-2-(4-methoxybenzoyl)-1-benzofuran

  • MDL number MFCD12922712
  • Cas Number : 220526-76-1
  • Molecular Weight : 286.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136 - 137
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101000

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KD-0736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart